3-Bromo-4-methyl-5-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific

PC303601-1G PC303601-5G
APOSPC303601-1GEA 0 EUR
APOSPC303601-1G APOSPC303601-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl bromide 97%
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl bromide
Formel: C₉H₇Br₂F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD32640174
CAS-Nummer: 2385223-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR