3-Bromo-4-methyl-5-(trifluoromethyl)benzyl bromide 97%
Lieferant: Apollo Scientific
PC303601-1G
PC303601-5G
APOSPC303601-1GEA
348
EUR
APOSPC303601-1G
APOSPC303601-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl bromide 97%
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl bromide
Formel:
C₉H₇Br₂F₃ Molecular Weight: 331.955 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640174 CAS-Nummer: 2385223-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...