3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetic acid

Lieferant: Apollo Scientific

PC303603-1G
APOSPC303603-1GEA 0 EUR
APOSPC303603-1G
3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetic acid
3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetic acid
Formel: C₁₀H₈BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD32640176
CAS-Nummer: 2387022-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR