3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
PC303605-1G
PC303605-5G
APOSPC303605-1GEA
279
EUR
APOSPC303605-1G
APOSPC303605-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid
Formel:
C₉H₆BrF₃O₂ Molecular Weight: 283.042 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640178 CAS-Nummer: 2324179-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...