3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303605-1G PC303605-5G
APOSPC303605-1GEA 0 EUR
APOSPC303605-1G APOSPC303605-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-4-methyl-5-(trifluoromethyl)benzoic acid
Formel: C₉H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD32640178
CAS-Nummer: 2324179-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR