3-Bromo-4-methyl-5-(trifluoromethyl)aniline 98%
Lieferant: Apollo Scientific
PC303609-1G
PC303609-5G
APOSPC303609-1GEA
430
EUR
APOSPC303609-1G
APOSPC303609-5G
3-Bromo-4-methyl-5-(trifluoromethyl)aniline 98%
3-Bromo-4-methyl-5-(trifluoromethyl)aniline
Formel:
C₈H₇BrF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD28746059 CAS-Nummer: 1219948-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...