3-Bromo-4-methyl-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303610-1G
APOSPC303610-1GEA 1050 EUR
APOSPC303610-1G
3-Bromo-4-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
3-Bromo-4-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₈H₅BrClF₃O₂S
Molecular Weight: 337,541 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640182
CAS-Nummer: 2383371-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR