6-Ethoxy-2,3-difluorobenzonitrile 97%

Lieferant: Apollo Scientific

PC303611-1G
APOSPC303611-1GEA 434 EUR
APOSPC303611-1G
6-Ethoxy-2,3-difluorobenzonitrile 97%
6-Ethoxy-2,3-difluorobenzonitrile
Formel: C₉H₇F₂NO
Molecular Weight: 183,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28765048
CAS-Nummer: 1807084-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR