3-Chloro-2-fluoro-5-(trifluoromethoxy)benzoic acid 97%

Lieferant: Apollo Scientific

PC303613-1G PC303613-250MG
APOSPC303613-1GEA 511 EUR
APOSPC303613-1G APOSPC303613-250MG
3-Chloro-2-fluoro-5-(trifluoromethoxy)benzoic acid 97%
3-Chloro-2-fluoro-5-(trifluoromethoxy)benzoic acid
Formel: C₈H₃ClF₄O₃
Molecular Weight: 258.554 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32689357
CAS-Nummer: 2385608-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR