Sanger's Reagenz 99%

Lieferant: Apollo Scientific
Gefahr

PC3040-100G PC3040-1KG PC3040-500G
APOSPC3040-100GEA 0 EUR
APOSPC3040-100G APOSPC3040-1KG APOSPC3040-500G
Sanger's Reagenz 99%
Sanger's Reagenz

Nucleophiles can displace the activated fluorine atom.Reagent for labelling of terminal amino acid groups.

Formel: C₆H₃FN₂O₄
Schmelzpunkt: 28…30 °C
Dichte: 1,482 g/cm³ (25 °C)
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007056
CAS-Nummer: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR