5-(Trifluoromethyl)indole
Lieferant: Apollo Scientific
|
Achtung
|
PC3045-1G
APOSPC3045-1GEA
226
EUR
APOSPC3045-1G
5-(Trifluoromethyl)indole
5-(Trifluoromethyl)indole
Formel:
C₉H₆F₃N Molecular Weight: 185,15 g/mol Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095341 CAS-Nummer: 100846-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...