4-(2,4-Difluorobenzoyl)piperidine hydrochloride

Lieferant: Apollo Scientific

PC305000-100G PC305000-25G
APOSPC305000-100GEA 185 EUR
APOSPC305000-100G APOSPC305000-25G
4-(2,4-Difluorobenzoyl)piperidine hydrochloride
4-(2,4-Difluorobenzoyl)piperidine hydrochloride
Formel: C₁₂H₁₃F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD01313310
CAS-Nummer: 106266-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR