1,2,3-Trifluoro-5-methoxy-4-nitrobenzene 98%

Lieferant: Apollo Scientific

PC305001-5G
APOSPC305001-5GEA 56.7 EUR
APOSPC305001-5G
1,2,3-Trifluoro-5-methoxy-4-nitrobenzene 98%
1,2,3-Trifluoro-5-methoxy-4-nitrobenzene
Formel: C₇H₄F₃NO₃
Molecular Weight: 207,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025341
CAS-Nummer: 925890-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR