3-(Trifluoromethyl)azetidin-3-ol hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

PC305004-1G PC305004-250MG
APOSPC305004-1GEA 90.5 EUR
APOSPC305004-1G APOSPC305004-250MG
3-(Trifluoromethyl)azetidin-3-ol hydrochloride 98%
3-(Trifluoromethyl)azetidin-3-ol hydrochloride
Formel: C₄H₇ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13248564
CAS-Nummer: 848192-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR