2-Amino-4-(trifluoromethyl)benzamide 98%

Lieferant: Apollo Scientific

PC305010-250MG
APOSPC305010-250MGEA 0 EUR
APOSPC305010-250MG
2-Amino-4-(trifluoromethyl)benzamide 98%
2-Amino-4-(trifluoromethyl)benzamide
Formel: C₈H₇F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06808543
CAS-Nummer: 713-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR