6-Bromo-5-fluoropyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-fluoropicolinic acid

PC305018-5G
APOSPC305018-5GEA 81.2 EUR
APOSPC305018-5G
6-Bromo-5-fluoropyridine-2-carboxylic acid
6-Bromo-5-fluoropyridine-2-carboxylic acid
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13185797
CAS-Nummer: 1052714-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR