4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC305027-250MG PC305027-1G
APOSPC305027-250MGEA 139 EUR
APOSPC305027-250MG APOSPC305027-1G
4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrF₄O₂
MDL: MFCD31695851
CAS-Nummer: 2092502-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR