4-Methoxy-2-(trifluormethyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3068-250MG
APOSPC3068-250MGEA
0
EUR
APOSPC3068-250MG
4-Methoxy-2-(trifluormethyl)phenylboronsäure 98%
4-Methoxy-2-(trifluormethyl)phenylboronsäure
Formel:
C₈H₈BF₃O₃ Schmelzpunkt: 168…172 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03095346 CAS-Nummer: 313546-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...