3-(Difluoromethoxy)phenacyl bromide

Lieferant: Apollo Scientific
Gefahr

PC3073-250MG PC3073-5G PC3073-1G
APOSPC3073-250MGEA 0 EUR
APOSPC3073-250MG APOSPC3073-5G APOSPC3073-1G
3-(Difluoromethoxy)phenacyl bromide
3-(Difluoromethoxy)phenacyl bromide
Formel: C₉H₇BrF₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09259003
CAS-Nummer: 443914-96-3
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR