2-Methoxy-4-(trifluormethyl)phenylboronsäure

Lieferant: Apollo Scientific

Synonyme: (2-Methoxy-4-(trifluoromethyl)phenyl)boronic acid

PC3077-5G PC3077-10G PC3077-1G
APOSPC3077-5GEA 93.4 EUR
APOSPC3077-5G APOSPC3077-10G APOSPC3077-1G
2-Methoxy-4-(trifluormethyl)phenylboronsäure
2-Methoxy-4-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219.9500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428511
CAS-Nummer: 312936-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR