2-Fluor-3-(trifluormethyl)zimtsäure

Lieferant: Apollo Scientific
Achtung

PC3079-1G PC3079-5G
APOSPC3079-1GEA 21.9 EUR
APOSPC3079-1G APOSPC3079-5G
2-Fluor-3-(trifluormethyl)zimtsäure
2-Fluor-3-(trifluormethyl)zimtsäure
Formel: C₁₀H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00236291
CAS-Nummer: 237069-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR