3,3'-Bis(trifluormethyl)benzhydrol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3086D-1G
APOSPC3086D-1GEA
157
EUR
APOSPC3086D-1G
3,3'-Bis(trifluormethyl)benzhydrol 97%
3,3'-Bis(trifluormethyl)benzhydrol
Formel:
C₁₅H₁₀F₆O Siedepunkt: 118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039228 CAS-Nummer: 1598-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...