3,5-Bis(trifluormethyl)benzhydrazid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Bis(trifluoromethyl)benzohydrazide
PC3090C-100G
PC3090C-25G
APOSPC3090C-100GEA
401
EUR
APOSPC3090C-100G
APOSPC3090C-25G
3,5-Bis(trifluormethyl)benzhydrazid 97%
3,5-Bis(trifluormethyl)benzhydrazid
Formel:
C₉H₆F₆N₂O Schmelzpunkt: 132…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051848 CAS-Nummer: 26107-82-4 EINECS: 247-462-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...