2,4-Bis(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Gefahr

PC3095-1G PC3095-5G PC3095-25G
APOSPC3095-1GEA 0 EUR
APOSPC3095-1G APOSPC3095-5G APOSPC3095-25G
2,4-Bis(trifluormethyl)benzonitril 98%
2,4-Bis(trifluormethyl)benzonitril
Formel: C₉H₃F₆N
Lagertemperatur: Raumtemperatur
MDL: MFCD00061225
CAS-Nummer: 177952-38-4
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR