2,6-Bis(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Gefahr

PC3096-250MG
APOSPC3096-250MGEA 60.9 EUR
APOSPC3096-250MG
2,6-Bis(trifluormethyl)benzonitril 98%
2,6-Bis(trifluormethyl)benzonitril
Formel: C₉H₃F₆N
Molecular Weight: 239.1172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061226
CAS-Nummer: 25753-25-7
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR