2,4-Bis(trifluormethyl)benzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC3104F-500MG
APOSPC3104F-500MGEA
21.9
EUR
APOSPC3104F-500MG
2,4-Bis(trifluormethyl)benzylalkohol 98%
2,4-Bis(trifluormethyl)benzylalkohol
Formel:
C₉H₆F₆O Molecular Weight: 244.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042491 CAS-Nummer: 143158-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...