1-Brom-3,5-bis(trifluormethyl)benzol 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC3106-100G
PC3106-25G
PC3106-500G
APOSPC3106-100GEA
39.3
EUR
APOSPC3106-100G
APOSPC3106-25G
APOSPC3106-500G
1-Brom-3,5-bis(trifluormethyl)benzol 99%
1-Brom-3,5-bis(trifluormethyl)benzol
Reagent for adding inductively withdrawing substituent into precursors for carbocations, J.O.C., 1984, 49, 2608
Formel:
C₈H₃BrF₆ Siedepunkt: 155 °C Schmelzpunkt: –16 °C Dichte: 1,699 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000381 CAS-Nummer: 328-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...