3,5-Bis(trifluoromethyl)-N-ethylaniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC3107K-100MG
APOSPC3107K-100MGEA
98.6
EUR
APOSPC3107K-100MG
3,5-Bis(trifluoromethyl)-N-ethylaniline 97%
3,5-Bis(trifluoromethyl)-N-ethylaniline
Formel:
C₁₀H₉F₆N Siedepunkt: 96…98 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00203993 CAS-Nummer: 49850-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...