2-Benzoxazol-2-yl-1-[(3-trifluoromethyl)phenyl]ethanol 95+%

Lieferant: Apollo Scientific
Gefahr

PC3110-1G
APOSPC3110-1GEA 50.9 EUR
APOSPC3110-1G
2-Benzoxazol-2-yl-1-[(3-trifluoromethyl)phenyl]ethanol 95+%
2-Benzoxazol-2-yl-1-[(3-trifluoromethyl)phenyl]ethanol
Formel: C₁₆H₁₂F₃NO₂
Molecular Weight: 307,27 g/mol
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03424485
CAS-Nummer: 502625-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR