2-Brom-5-(trifluormethyl)anisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-3-methoxybenzotrifluoride

PC3113-25G
APOSPC3113-25GEA 148 EUR
APOSPC3113-25G
2-Brom-5-(trifluormethyl)anisol 98%
2-Brom-5-(trifluormethyl)anisol
Formel: C₈H₆BrF₃O
Molecular Weight: 255.03 g/mol
Schmelzpunkt: 26…28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115974
CAS-Nummer: 402-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR