1-(4-Bromo-2-fluorobenzyl)homopiperazine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(4-Bromo-2-fluorobenzyl)-1,4-diazepane

PC31133-1G
APOSPC31133-1GEA 173 EUR
APOSPC31133-1G
1-(4-Bromo-2-fluorobenzyl)homopiperazine
1-(4-Bromo-2-fluorobenzyl)homopiperazine
Formel: C₁₂H₁₆BrFN₂
Molecular Weight: 287,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682103
CAS-Nummer: 646455-62-1
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR