4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N,N-Dimethyl-2-(trifluoroacetyl)ethenylamine, N,N-Dimethyl-2-(trifluoroacetyl)vinylamine
PC31140-1G
PC31140-5G
APOSPC31140-1GEA
163
EUR
APOSPC31140-1G
APOSPC31140-5G
4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
Formel:
C₆H₈F₃NO Molecular Weight: 167.129 g/mol Schmelzpunkt: 56…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00764476 CAS-Nummer: 142503-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...