4-Amino-1,1,1-trifluorobut-3-en-2-one
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Amino-2-oxo-1,1,1-trifluorobut-3-ene, 4-Amino-1,1,1-trifluoro-3-buten-2-one
PC31145-1G
APOSPC31145-1GEA
87
EUR
APOSPC31145-1G
4-Amino-1,1,1-trifluorobut-3-en-2-one
4-Amino-1,1,1-trifluorobut-3-en-2-one
Formel:
C₄H₄F₃NO Molecular Weight: 139,076 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569588 CAS-Nummer: 184848-89-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...