1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-{[4-(pent-1-yl)phenyl]sulphonyl}homopiperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-2-(4-{[4-(pent-1-yl)phenyl]sulphonyl}-1,4-diazepan-1-yl)-5-(trifluoromethyl)pyridine, 1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-{[4-(pent-1-yl)phenyl]sulphonyl}-1,4-diazepane

PC31148-250MG
APOSPC31148-250MGEA 116 EUR
APOSPC31148-250MG
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-{[4-(pent-1-yl)phenyl]sulphonyl}homopiperazine
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-4-{[4-(pent-1-yl)phenyl]sulphonyl}homopiperazine
Formel: C₂₂H₂₇ClF₃N₃O₂S
Molecular Weight: 489,982 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179834
CAS-Nummer: 646456-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR