3,5-Bis(trifluoromethyl)-DL-phenylalanine 98%

Lieferant: Apollo Scientific
Achtung

PC3114L-1G PC3114L-500MG
APOSPC3114L-1GEA 46.4 EUR
APOSPC3114L-1G APOSPC3114L-500MG
3,5-Bis(trifluoromethyl)-DL-phenylalanine 98%
3,5-Bis(trifluoromethyl)-DL-phenylalanine
Formel: C₁₁H₉F₆NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD00061309
CAS-Nummer: 237076-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR