N1-(4-Fluorophenyl)-2-aminobenzamide

Lieferant: Apollo Scientific

Synonyme: 2-Amino-N-(4-fluorophenyl)benzamide

PC31159-250MG
APOSPC31159-250MGEA 55.3 EUR
APOSPC31159-250MG
N1-(4-Fluorophenyl)-2-aminobenzamide
N1-(4-Fluorophenyl)-2-aminobenzamide
Formel: C₁₃H₁₁FN₂O
Molecular Weight: 230,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00115471
CAS-Nummer: 216502-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR