1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3115M-1G
PC3115M-5G
APOSPC3115M-1GEA
61.1
EUR
APOSPC3115M-1G
APOSPC3115M-5G
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff 98%
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
Formel:
C₉H₆F₆N₂S Schmelzpunkt: 168…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041172 CAS-Nummer: 175277-17-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...