2'-Amino-4'-(trifluoromethyl)acetanilide
Lieferant: Apollo Scientific
|
Achtung
|
PC31160-100MG
PC31160-1G
PC31160-250MG
APOSPC31160-100MGEA
53.8
EUR
APOSPC31160-100MG
APOSPC31160-1G
APOSPC31160-250MG
2'-Amino-4'-(trifluoromethyl)acetanilide
2'-Amino-4'-(trifluoromethyl)acetanilide
Formel:
C₉H₉F₃N₂O Schmelzpunkt: 141…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018052 CAS-Nummer: 97051-69-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...