N-[(1,3-Benzodioxol-5-yl)methyl]-4-chloro-N-(4-fluorophenyl)-3-nitrobenzenesulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-({[(4-Chloro-3-nitrophenyl)sulphonyl](4-fluorophenyl)amino}methyl)-1,3-benzodioxole
PC31161-100MG
APOSPC31161-100MGEA
75.4
EUR
APOSPC31161-100MG
N-[(1,3-Benzodioxol-5-yl)methyl]-4-chloro-N-(4-fluorophenyl)-3-nitrobenzenesulphonamide
N-[(1,3-Benzodioxol-5-yl)methyl]-4-chloro-N-(4-fluorophenyl)-3-nitrobenzenesulphonamide
Formel:
C₂₀H₁₄ClFN₂O₆S Molecular Weight: 464,851 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00098183 CAS-Nummer: 216501-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...