N-(2-Chloro-6-fluoro-3-nitrophenyl)acetamide 95+%

Lieferant: Apollo Scientific

PC31203-250MG
APOSPC31203-250MGEA 258 EUR
APOSPC31203-250MG
N-(2-Chloro-6-fluoro-3-nitrophenyl)acetamide 95+%
N-(2-Chloro-6-fluoro-3-nitrophenyl)acetamide
Formel: C₈H₆ClFN₂O₃
Molecular Weight: 232,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00275692
CAS-Nummer: 218796-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR