2-Bromo-N-(5,7-difluoro-1,3-benzothiazol-2-yl)acetamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Bromoacetamido)-5,7-difluoro-1,3-benzothiazole, 2-[(Bromoacetyl)amino]-5,7-difluorobenzo[d]thiazole
PC31221-250MG
APOSPC31221-250MGEA
180
EUR
APOSPC31221-250MG
2-Bromo-N-(5,7-difluoro-1,3-benzothiazol-2-yl)acetamide
2-Bromo-N-(5,7-difluoro-1,3-benzothiazol-2-yl)acetamide
Formel:
C₉H₅BrF₂N₂OS Molecular Weight: 307,115 g/mol Schmelzpunkt: 191…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00177782 CAS-Nummer: 219499-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...