N1-(2,3,4-trifluorophenyl)-2-chloroacetamide
Lieferant: Apollo Scientific
PC31270-1G
APOSPC31270-1GEA
183
EUR
APOSPC31270-1G
N1-(2,3,4-trifluorophenyl)-2-chloroacetamide
N1-(2,3,4-trifluorophenyl)-2-chloroacetamide
Formel:
C₈H₅ClF₃NO Molecular Weight: 223,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00828901 CAS-Nummer: 243644-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...