7H-Perfluoroheptanoyl fluoride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC3131G-5G
APOSPC3131G-5GEA
0
EUR
APOSPC3131G-5G
7H-Perfluoroheptanoyl fluoride 97%
7H-Perfluoroheptanoyl fluoride
Formel:
C₇HF₁₃O Siedepunkt: 91 °C (630 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155774 CAS-Nummer: 5927-65-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...