4-Brom-3-(trifluormethyl)anisol 98%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-5-methoxybenzotrifluoride, 4-Bromo-3-trifluoromethylanisole
PC3133-25G
APOSPC3133-25GEA
74.1
EUR
APOSPC3133-25G
4-Brom-3-(trifluormethyl)anisol 98%
4-Brom-3-(trifluormethyl)anisol
Formel:
C₈H₆BrF₃O Molecular Weight: 255,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059527 CAS-Nummer: 400-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...