4-(Trifluormethyl)cyclohexanol (cis/trans-Gemisch)

Lieferant: Apollo Scientific
Achtung

PC31341-5G
APOSPC31341-5GEA 72.5 EUR
APOSPC31341-5G
4-(Trifluormethyl)cyclohexanol (cis/trans-Gemisch)
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)
Formel: C₇H₁₁F₃O
Siedepunkt: 70…75 °C/15mmHg
Dichte: 1,23 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00102144
CAS-Nummer: 30129-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR