5-Brom-2-fluorpyrimidin

Lieferant: Apollo Scientific
Achtung

PC3139-1G
APOSPC3139-1GEA 21.9 EUR
APOSPC3139-1G
5-Brom-2-fluorpyrimidin
5-Bromo-2-fluoropyrimidine
Formel: C₄H₂BrFN₂
Schmelzpunkt: 90…93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07787364
CAS-Nummer: 62802-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR