N1-(2,6-difluorophenyl)-2-chloroacetamide

Lieferant: Apollo Scientific

PC31468-1G
APOSPC31468-1GEA 226 EUR
APOSPC31468-1G
N1-(2,6-difluorophenyl)-2-chloroacetamide
N1-(2,6-difluorophenyl)-2-chloroacetamide
Formel: C₈H₆ClF₂NO
Molecular Weight: 205,589 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00119514
CAS-Nummer: 19064-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR