Fluorcyclopentan

Lieferant: Apollo Scientific
Achtung

PC3147-25G PC3147-5G
APOSPC3147-25GEA 693 EUR
APOSPC3147-25G APOSPC3147-5G
Fluorcyclopentan
Fluorocyclopentane
Formel: C₅H₉F
Lagertemperatur: Raumtemperatur
MDL: MFCD08059532
CAS-Nummer: 1481-36-3
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR