N1-(4-bromo-2,6-difluorophenyl)-2-chloroacetamide
Lieferant: Apollo Scientific
PC31501-1G
APOSPC31501-1GEA
183
EUR
APOSPC31501-1G
N1-(4-bromo-2,6-difluorophenyl)-2-chloroacetamide
N1-(4-bromo-2,6-difluorophenyl)-2-chloroacetamide
Formel:
C₈H₅BrClF₂NO Molecular Weight: 284,485 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00177966 CAS-Nummer: 646989-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...