3-Brom-4-nitrobenzotrifluorid 97%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-1-nitro-4-(trifluoromethyl)benzene
PC3153-5G
APOSPC3153-5GEA
121
EUR
APOSPC3153-5G
3-Brom-4-nitrobenzotrifluorid 97%
3-Brom-4-nitrobenzotrifluorid
Formel:
C₇H₃BrF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998160 CAS-Nummer: 132839-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...